Bromophenol blue solution R1 Reag. Ph. Eur. 1012802
Catalog # 87795.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:669.97 g/mol
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875