Order Entry
Austria
ContactUsLinkComponent
Bromophenol blue solution Reag. Ph. Eur. 1012801
Bromophenol blue solution Reag. Ph. Eur. 1012801
Catalog # 87794.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromophenol blue solution Reag. Ph. Eur. 1012801
Catalog # 87794.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :669.97 g/mol
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875