
Palladium(II) nitrate 10.0 ± 0.2 g/L in nitric acid 15% matrix modifier solution, for graphite furnace AAS, Supelco®
Catalog # 1.07289.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Assay (palladium):9.8 - 10.2 g/l
- Ag (Silver):≤ 0.000020 g/l
- As (Arsenic):≤ 0.000010 g/l
- Cd (Cadmium):≤ 0.000001 g/l
- Pb (Lead):≤ 0.000010 g/l
- Se (Selenium):≤ 0.00010 g/l
- Tl (Thallium):≤ 0.000020 g/l
Specification Test Results
About this item
:
:230.43 g/mol
Density:1.11 g/cm3 (20 °C)
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:2031
:GHS05
CAS Number:10102-05-3
Formula:N₂O₆Pd
MDL Number:MFCD00011169