Order Entry
Austria
ContactUsLinkComponent
 
Granular dessicant, Sicapent® with indicator, Supelco®
  1.00543.0500
 :  Merck
Granular dessicant, Sicapent® with indicator, Supelco®
  1.00543.0500
 :  Merck

 

  • Particle Size:
    0,1 - 1,6 mm
  • Description:
    SICAPENT® with indicator
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ200
  • Pk:
    500 mL

 

 

Sicapent is a granular dessicant comprising ~75% phoshorus pentoxide on an inert inorganic carrier.

  • Ease of use
  • Fast water adsorption due to large surface area
  • Colour indicator - colourless to green to blue
  • Remains in particle form after 100% water uptake

Appliations include drying liquids, filling drying tubes, particularly for fast flowing gases in drying tubes.

Bulk density: ~300 g/L