Order Entry
Austria
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for gas chromatography, Supelco®
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for gas chromatography, Supelco®
Catalog # 1.11805.0005
Supplier:  Merck
CAS Number:  
Flammable
Irritant
N-Methyl-N-(trimethylsilyl)trifluoroacetamide for gas chromatography, Supelco®
Catalog # 1.11805.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Molar Mass:
    199.25 g/mol
  • Boiling Point:
    130 - 132 °C (1013 hPa)
  • Flash Point:
    25 °C
  • Density:
    1.085 g/cm3
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

CAS Number:24589-78-4
 :
Formula:C₆H₁₂F₃NOSi
MDL Number:MFCD00000411
 :199.25 g/mol
Boiling Pt:130 °C (1013 hPa)
Density:1.07 g/cm³ (20 °C)
EINECS:246-331-6
UN:1993
ADR:3,
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS07
Storage Temperature:Ambient