Orders LinkContactUsLinkComponent
Infusion filters set, Intrapur® Inline
Infusion filters set, Intrapur® Inline
Infusion sets for use with chemotherapeutic agents, 0,2 µm
Catalog # BRAU4099842N
Supplier:   B. Braun
Infusion filters set, Intrapur® Inline
Catalog # BRAU4099842N
Supplier:  B. Braun
Infusion filters set, Intrapur® Inline
Infusion filters set, Intrapur® Inline
Infusion sets for use with chemotherapeutic agents, 0,2 µm
Price
682,00 €
Log in for availability
1 * 20 ST
Product Details & Documents
Ready-to-use set with infusion filter.
  • PVC free
  • Retention of bacteria and particles
  • High throughput
Specifically designed for use during chemotherapy.
Specifications
  • Membrane:
    0,2 µm
  • Description:
    Infusion sets for use with chemotherapeutic agents