Order Entry
Austria
ContactUsLinkComponent
Insulin syringes, Omnifix® 40 Solo
Insulin syringes, Omnifix® 40 Solo
Catalog # BRAU9161309V
Supplier:  B. Braun
undefined
Insulin syringes, Omnifix® 40 Solo
Catalog # BRAU9161309V
Supplier:  B. Braun

Specifications

  • Capacity:
    1 ml
  • Description:
    Omnifix® 40 Solo without needle, centric Luer cone, for U-40 insulin (1 ml/40 I.U.)
  • Environmentally Preferable:
  • Graduation:
    0,025 ml
  • Pk:
    100

Specifications

About this item

Omnifix® syringes are single-use insulin syringes for U-40.

  • Latex-free plug
  • Safe piston stop avoids loss of medicine
  • Highly transparent barrel