Orders LinkContactUsLinkComponent
Cell production roller apparatus, modular
Cell production roller apparatus, modular
Marketsource
Supplier:   BELLCO GLASS
Cell production roller apparatus, modular
Supplier:  BELLCO GLASS
Marketsource
Certificates
Cell production roller apparatus, modular
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Availability
    Price
  • BELC7630-75222
    Base with 2 decks, 15 bottle positions, 750×630×730 mm
    Base with 2 decks, 15 bottle positions, 750×630×730 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11 680,00 €
    11 680,00 €
    1 * 1 ST
       11 680,00 €
    Specifications:
    Marketsource
    Marketsource
  • BELC7630-75224
    Base with 4 decks, 25 bottle positions, 750×630×1090 mm
    Base with 4 decks, 25 bottle positions, 750×630×1090 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16 040,00 €
    16 040,00 €
    1 * 1 ST
       16 040,00 €
    Specifications:
    Marketsource
    Marketsource
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • All-position drive conversion kit
    All-position drive conversion kit
    BELC7630-50555
    Log in for availability
    1 * 1 ST - 302,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 5-position deck
    5-position deck
    BELC7630-75010
    Log in for availability
    1 * 1 ST - 2 340,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 5-position base
    5-position base
    BELC7630-75220
    Log in for availability
    1 * 1 ST - 7 320,00 €