Sterilisation indicator tapes
: DELTALAB
Sterilisation indicator tapes for steam autoclave.
According to models tape has printed on it as CONTAMINATED or batch, con. (control) and date. After sterilisation to +121 °C the word STERILISED appears.
Frequently Bought Together