Water sampling bottles, rectangular
About this item
Polyethylene, translucent, with red screw cap.
- Tamper evident red cap with an internal joint
- Hermetical closure
- Sterilised by radiation
- Available with or without sodium thiosulfate
With liquid sodium thiosulphate (24 mg/L): ideal to analyse water meant for human consumption, swimming pools, and any other water where chlorine may modify the composition of the sample while being transported.
Without sodium thiosulphate: ideal to sample water for physico-chemical analysis, as well as for other sampling requiring sterile bottles.
The minor sides are gripped for a better handling. The larger sides are flat for labelling. Each bottle is labelled specifying description, code, lot, and expiry date.
8 Options Available Below
Product Details & Documents
Polyethylene, translucent, with red screw cap.
- Tamper evident red cap with an internal joint
- Hermetical closure
- Sterilised by radiation
- Available with or without sodium thiosulfate
With liquid sodium thiosulphate (24 mg/L): ideal to analyse water meant for human consumption, swimming pools, and any other water where chlorine may modify the composition of the sample while being transported.
Without sodium thiosulphate: ideal to sample water for physico-chemical analysis, as well as for other sampling requiring sterile bottles.
The minor sides are gripped for a better handling. The larger sides are flat for labelling. Each bottle is labelled specifying description, code, lot, and expiry date.
Documents
Product Family Options
Water sampling bottles with thiosulphate
Product Information
- CapacityDosageCap colourWithW×D×HNeck Ø ext.PackedAvailabilityPrice
- DELT282320Bottle with thiosulfateBottle with thiosulfate500 mlRedLabel83×65×135 mm28 mm24/caseLog in for availability1 * 24 ST92,70 €
- DELT282321Bottle with thiosulfateBottle with thiosulfate1000 mlRedLabel83×65×235 mm28 mm20/caseLog in for availability1 * 20 ST91,30 €
- DELT282323Bottle with thiosulfateBottle with thiosulfate500 mlRedLabel83×65×135 mm28 mm111/caseLog in for availability1 * 111 ST389,00 €
- DELT282323.BUBottle with thiosulfate, individually wrappedBottle with thiosulfate, individually wrapped500 mlRedLabel83×65×135 mm28 mm111/caseLog in for availability1 * 111 ST472,00 €
- DELT282324Bottle with thiosulfateBottle with thiosulfate1000 mlRedLabel83×65×235 mm28 mm68/caseLog in for availability1 * 68 ST287,00 €
- DELT282324.BUBottle with thiosulfate, individually wrappedBottle with thiosulfate, individually wrapped1000 mlRedLabel83×65×235 mm28 mm68/caseLog in for availability1 * 68 ST338,00 €
Water sampling bottles without thiosulphate
Product Information
- CapacityDosageCap colourWithW×D×HNeck Ø ext.PackedAvailabilityPrice
- DELT282330Bottle without thiosulfateBottle without thiosulfate500 mlUndosedRedLabel83×65×135 mm28 mm24/caseLog in for availability1 * 24 ST90,20 €
- DELT282331Bottle without thiosulfateBottle without thiosulfate1000 mlUndosedRedLabel83×65×235 mm28 mm20/caseLog in for availability1 * 20 ST91,20 €