ContactUsLinkComponent
Soxhlet extractors
Soxhlet extractors
Catalog # KAVA632442101100
Supplier:  Kavalier
undefined
Soxhlet extractors
Catalog # KAVA632442101100
Supplier:  Kavalier
Specifications
  • Extraction cap.:
    100 ml
  • Socket size:
    45/40 NS
  • Description:
    Extractor with cone 29/32
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Borosilicate glass. For extraction from solids with solvents.
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.