Orders LinkContactUsLinkComponent
Separating funnel, globe shaped
Separating funnel, globe shaped
Supplier:   Kavalier
Separating funnel, globe shaped
Supplier:  Kavalier
Certificates
Separating funnel, globe shaped
Separating funnel, globe shaped
2 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for separation of particular phases of the mixture. With standard ground joint stopper.
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 306,00 €
    306,00 €
    1 * 1 ST
       306,00 €
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 451,00 €
    451,00 €
    1 * 2 ST
       451,00 €