Order Entry
Austria
ContactUsLinkComponent
Test tubes, round bottom, without rim
Test tubes, round bottom, without rim
Catalog # KAVA632422020180
Supplier:  Kavalier
Test tubes, round bottom, without rim
Catalog # KAVA632422020180
Supplier:  Kavalier

Specifications

  • Ext.Ø:
    20 mm
  • Wall thickness:
    1,2 mm
  • Length:
    180 mm
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes