Orders LinkContactUsLinkComponent
Extraction unit and drying attachment, LiChrolut®
Supplier:   Merck
Extraction unit and drying attachment, LiChrolut®
Supplier:  Merck
Certificates
Extraction unit and drying attachment, LiChrolut®
Extraction unit and drying attachment, LiChrolut®
3 Options Available Below
Product Details & Documents
All the individual steps associated with solid phase extraction can be carried out using the LiChrolut® extraction unit. This transparent and vacuum-suitable unit, made of glass, can be used to prepare up to 12 samples simultaneously.
  • Control of the vacuum via a manometer located at the front
  • Individual, easy setting of the various flow rates using valves
 : Supplied with 1 lid with 12 standard valves and seal, 1 glass chamber with gauge and vacuum valve, 12 standard stainless steel cannules, 1 collecting rack (base plate with 3 support rods, centre plate, top plate with 10 mm boring and 12 clamps), 1 rack for volumetric flasks, 1 rack for 16 mm test tubes and 1 rack for autosampler vials.
Product Family Options
Product Information
  • Availability
    Price
  • 1.19851.0001
    Complete extraction unit, LiChrolut®
    Complete extraction unit, LiChrolut®
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 450,00 €
    2 450,00 €
    1 * 1 ST
       2 450,00 €
  • 1.19852.0001
    Complete drying attachment, LiChrolut®
    Complete drying attachment, LiChrolut®
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 630,00 €
    1 630,00 €
    1 * 1 ST
       1 630,00 €
  • 1.19874.0001
    Disposable fluoroplastic liners
    Disposable fluoroplastic liners
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 230,00 €
    230,00 €
    1 * 100 ST
       230,00 €