ContactUsLinkComponent
Dishes with lid
Dishes with lid
Catalog # NICK503543
Supplier:  NICKEL ELECTRO
undefined
Dishes with lid
Catalog # NICK503543
Supplier:  NICKEL ELECTRO
Specifications
  • Ext.Ø:
    60 mm
  • Version:
    flat form
  • Height:
    16 mm
  • Pk:
    1
Specifications
About this item
Made of nickel.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean