Order Entry
Austria
ContactUsLinkComponent
Carboys, with handles and stopcock, HDPE
Carboys, with handles and stopcock, HDPE
Catalog # 215-0290
Supplier:  SEMADENI
undefined
Carboys, with handles and stopcock, HDPE
Catalog # 215-0290
Supplier:  SEMADENI

Specifications

  • Capacity:
    10 l
  • W×D×H:
    220×175×390 mm
  • Neck-Ø:
    63 mm
  • Pk:
    1

Specifications

About this item

HDPE, translucent.

  • Good chemical resistance
  • Good temperature resistance

With stopcock, black screw cap and black handle.