ContactUsLinkComponent
Sieve homogenisers, Cellector™
Sieve homogenisers, Cellector™
Catalog # BELC1985-85000
Supplier:  BELLCO GLASS
Sieve homogenisers, Cellector™
Catalog # BELC1985-85000
Supplier:  BELLCO GLASS
Specifications
  • Description:
    Cellector™, complete, 85 ml (pan)
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Cellector™ homogenisers, made of stainless steel, are for preparation of cell suspensions and gel extraction of proteins.
Delivery: Cellector™ complete includes a pan, glass pestle, key and nine screens (one each) 10 to 150 mesh size.