Order Entry
Austria
ContactUsLinkComponent
 
Sodium molybdate(VI) dihydrate ≥98.0% (by titration analysis on dried basis) for synthesis, Sigma-Aldrich®
 :  Merck
 
Sodium molybdate(VI) dihydrate ≥98.0% (by titration analysis on dried basis) for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Hill Formula:
    Na₂MoO₄ * 2 H₂O
  • Chemical formula:
    Na₂MoO₄ * 2 H₂O
  • Molar Mass:
    241.95 g/mol
  • pH value:
    9 - 10 (840 g/l, H₂O, 20 °C)
  • Bulk density:
    1000 - 1400 kg/m³
  • Solubility:
    840 g/l (20 °C)
  • LD 50 oral:
    LD50 Rat 4233 mg/kg
  • LD 50 dermal:
    LD50 Rat 2000 mg/kg
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 1 slightly water endangering

 

 

Frequently Bought Together