ContactUsLinkComponent
Cylinders, mutipurpose
Cylinders, mutipurpose
Catalog # WITG5.630.004
Supplier:  witeg Labortechnik
undefined
Cylinders, mutipurpose
Catalog # WITG5.630.004
Supplier:  witeg Labortechnik
Specifications
  • Ø:
    40 mm
  • Height:
    200 mm
  • Pk:
    1
Specifications
About this item
Cylinders, made of DURAN® borosilicate glass, with rolled rim and round base.
  • Very good chemical resistance
  • High resistance to temperature change
  • No graduations