ContactUsLinkComponent
Fluid aspiration system, Biochem VacuuCenter, BVC control
undefined
Fluid aspiration system, Biochem VacuuCenter, BVC control
  • Catalog No:
  • 181-0293
  • 181-0321
  • Model:
  • BioChem-VacuuCenter BVC control
  • BioChem-VacuuCenter BVC control G
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Vacuum pump:
  • ME 1C
  • ME 1C
  • Final pressure of vacuum pump:
  • 150 mbar
  • 150 mbar
  • Protection class:
  • IP 40
  • IP 40
  • Weight:
  • 7,3 kg
  • 7,3 kg
Frequently Bought Together