Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
Supplier:   witeg Labortechnik
Double spatulas, micro
Supplier:  witeg Labortechnik
Certificates
Double spatulas, micro
Double spatulas, micro
2 Options Available Below
Product Details & Documents
Micro spatulas with two flat ends, made of stainless steel.
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Availability
    Price
  • 130 mm
    40 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,70 €
    1,70 €
    1 * 1 ST
       1,70 €
  • 150 mm
    40 mm
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,70 €
    1,70 €
    1 * 1 ST
       1,70 €