Safety wash bottles, narrow neck, with printed name
Supplier: witeg Labortechnik
Certificates
About this item
LDPE, yellow, with coloured screw caps.
- With hazard symbols and descriptions of hazards as required by the hazardous substance ordinance
- Good chemical resistance
- Good thermal resistance
With safety delivery jet.
Frequently Bought Together