Order Entry
Austria
ContactUsLinkComponent
Test tubes, PS, with screw caps
Test tubes, PS, with screw caps
Catalog # WITG5.464.102
Supplier:  witeg Labortechnik
undefined
Test tubes, PS, with screw caps
Catalog # WITG5.464.102
Supplier:  witeg Labortechnik

Specifications

  • Ext.Ø:
    16 mm
  • Capacity:
    10 ml
  • Length:
    100 mm
  • Pk:
    200

Specifications

About this item

PS, clear, with screw cap.

  • Sterile
  • Round bottom
  • Good thermal resistance
  • Good chemical resistance

Universal tubes for a wide range of applications.