Dropping bottles, glass, with ground-in stoppers
Supplier: Kavalier
Certificates
About this item
Glass, clear or brown.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
Amber to protect light-sensitive substances.
Frequently Bought Together