ContactUsLinkComponent
Repetitive pipettes, mechanical, DISTRIMAN®
Repetitive pipettes, mechanical, DISTRIMAN®
Catalog # 612-7810
Supplier:  GILSON
undefined
Repetitive pipettes, mechanical, DISTRIMAN®
Catalog # 612-7810
Supplier:  GILSON
Specifications
  • Capacity:
    1 - 1250 µl
  • Description:
    Manual repetitive pipette, DISTRIMAN®
  • Pk:
    1
Specifications
About this item
DISTRIMAN® is a positive-displacement, continuously adjustable repeater-pipette to simplify multiple dispensing when used with Gilson DISTRITIP® syringes.
  • Zero risk of accidental conversion errors with the direct read out
  • Time saving: continuous adjustable volume and numerous distribution of aliquots (up to 125)
  • Faster multiple dispensing
  • Control button for volume selection
  • DISTRITIP® syringes are available in standard and sterilised versions
DISTRIMAN® can dispense aliquots ranging in volume from 1 µl to 1,25 ml with the combination of its three dedicated models of DISTRITIP®. Each syringe can dispense up to 125 aliquots (of the minimal volume).
Ideal for any application requiring repetitive dispensing in the clinical, hospital, biological, chemical, food & beverage, pharmaceutical, cosmetics and forensics fields.
Syringe tips
Product Information