ContactUsLinkComponent
Crucible for determination of volatile liquids
  KAVA641321098000
 :  Kavalier
Crucible for determination of volatile liquids
  KAVA641321098000
 :  Kavalier
  • Ø×H:
    32×42 mm
  • Type:
    with lid, for determination of volatile liquid
  • Capacity:
    22 ml
  • Environmentally Preferable:
  • Pk:
    1
This crucible is made from porcelain.
 : Supplied with lid.