Orders LinkContactUsLinkComponent
Mops, PowerHead™
Mops, PowerHead™
Supplier:   MICRONOVA
Mops, PowerHead™
Supplier:  MICRONOVA
Certificates
Mops, PowerHead™
Mops, PowerHead™
4 Options Available Below
Product Details & Documents
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
  • Stainless steel PowerHead™ handles hold and wring the various mop heads
  • Autoclavable
  • A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Ordering information: All components are ordered separately.
Product Family Options
Product Information
  • Availability
    Price
  • 129-0523
    PowerHead™ absorbent foam,356 mm
    PowerHead™ absorbent foam,356 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,70 €
    35,70 €
    1 * 1 ST
       35,70 €
  • 129-0525
    PowerHead™ wide pore foam, 356 mm
    PowerHead™ wide pore foam, 356 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,60 €
    24,60 €
    1 * 1 ST
       24,60 €
  • MICRPH6-150
    PowerHead™ NovaPoly™ covered foam, 254 mm
    PowerHead™ NovaPoly™ covered foam, 254 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,00 €
    47,00 €
    1 * 1 ST
       47,00 €
  • 129-0521
    PowerHead™ NovaPoly™ covered foam, 356 mm
    PowerHead™ NovaPoly™ covered foam, 356 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,00 €
    47,00 €
    1 * 1 ST
       47,00 €