Order Entry
Austria
ContactUsLinkComponent
 
Mag-Bind® Plant DNA plus kit
 
Mag-Bind® Plant DNA plus kit

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Sample size:
    50 mg of wet tissue or 10 mg dry tissue
  • Description:
    Mag-Bind® Plant DNA Plus Kit

 

Frequently Bought Together