Order Entry
Austria
ContactUsLinkComponent
Fungal RNA kit, E.Z.N.A.®
Fungal RNA kit, E.Z.N.A.®
Supplier:  OMEGA BIO-TEK
Certificates
Fungal RNA kit, E.Z.N.A.®
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    RNA
  • Sample size:
    100 mg
  • Description:
    E.Z.N.A.® Fungal RNA Kit

Specifications

Related Information

Frequently Bought Together