Order Entry
Austria
ContactUsLinkComponent
RNA isolation, total RNA kit II, E.Z.N.A.®
RNA isolation, total RNA kit II, E.Z.N.A.®
Supplier:  OMEGA BIO-TEK
Certificates
RNA isolation, total RNA kit II, E.Z.N.A.®
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
    Organic extraction
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Fatty tissues
    Cultured cells
  • Target molecule:
    RNA
  • Sample size:
    10⁷ eukaryotic cells, up to 10⁹ bacterial cells, or 100 mg tissue
  • Description:
    E.Z.N.A.® Total RNA Kit II

Specifications

Related Information

Frequently Bought Together