Regenerating hand and body cream, Stokolan® sensitive
115-1743
- Description:Regenerating hand and body cream, Stokolan® Sensitive PURE, 1000 ml soft bottle for Stoko® Vario® dispensers
- Seq:02
- Environmentally Preferable:
- Pk:9
White soft cream (O/W emulsion) highly suitable for sensitive skin and for facial care. Dermatologically confirmed.
- With 9,5% NMF (glycerine and urea), creatine and other caring and regenerating substances
- Easy to apply
- Perfume-free, without preservatives, and free from colourants, silicone and parabens
: ECARF quality seal for allergy friendliness, HACCP.
: Soft bottle may only be used with Stoko® Vario® dispensers.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904