Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Supplier:   SEMADENI
Storage Boxes
Supplier:  SEMADENI
Certificates
Storage Boxes

2 Options Available Below

Product Details & Documents

These assortment and bearing boxes made of polystyrene (PS) are transparent with hinged lid and multiple longitudinal and transverse divisions.

Product Family Options

Product Information

  • Colour
    W×D×H
    Material
    Availability
    Price
  • 216-7311
    with hinged lid and 3×6 number of compartments
    with hinged lid and 3×6 number of compartments
    Transparent
    210×120×35 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,50 €
    27,50 €
    1 * 1 ST
       27,50 €

  • 216-7312
    with hinged lid and 4×6 number of compartments
    with hinged lid and 4×6 number of compartments
    Transparent
    330×220×53 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,10 €
    44,10 €
    1 * 1 ST
       44,10 €