Bovine desoxyribonuclease I Typ II (from Pancreas)
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:9003-98-9
- Source:Pancreas
- Species:Bovine
- Storage conditions:Freezer
- Enzyme name:Desoxyribonuclease I, Typ II
- Enzyme synonyms:DNase I
Specifications
Frequently Bought Together