Ammonium dihydrogen phosphate ≥98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7722-76-1
:
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396
Merck Index:13,00547
Melting Pt:190 °C
Density:1.80 g/cm³ (20 °C)
:115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥98.0%
- For Laboratory,Research,or Manufacturing Use:≤0.001%
- Assay (NH₄H₂PO₄):≥98.0 %
- Calcium (Ca):≤0.001 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:3.8 - 4.4
- Magnesium (Mg):≤0.0005 %
- Nitrate (NO₃):≤0.001 %
- Sulfate (SO₄):≤0.01 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.005 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together