Order Entry
Austria
ContactUsLinkComponent
Sodium peroxide ≥93.0%, granules, AR® ACS, Macron Fine Chemicals™
Sodium peroxide ≥93.0%, granules, AR® ACS, Macron Fine Chemicals™
Catalog # MACR7864-04
CAS Number:  
undefined
Sodium peroxide ≥93.0%, granules, AR® ACS, Macron Fine Chemicals™
Catalog # MACR7864-04
CAS Number:  

Specifications

  • Pack type:
    Metal can
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • ACS - Assay (Na₂O₂) (by KMnO₄ titrn):
    ≥93.0 %
  • ACS - Chloride (Cl):
    ≤0.002 %
  • ACS - Heavy Metals (as Pb):
    ≤0.002 %
  • ACS - Iron (Fe):
    ≤0.005 %
  • ACS - Phosphate (PO₄):
    ≤0.0005 %
  • ACS - Sulfate (SO₄):
    ≤0.001 %

Specification Test Results

About this item

Synonyms: di-Sodium peroxide
 :
 :77.98 g/mol
Merck Index:13,08728
Melting Pt:660 °C
Density:2.805 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-209-4
UN:1504
ADR:5.1,I
 :GHS03
 :GHS05
CAS Number:1313-60-6
Formula:Na₂O₂
MDL Number:MFCD00003497