Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
1,4-Dioxane ≥99.0% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
 
 
 
1,4-Dioxane ≥99.0% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (C₄H₈O₂) (by GC, corrected for water):
    ≥99.0 %
  • Carbonyl Compounds (as HCHO) (by polarography):
    ≤0.01 %
  • Color (APHA):
    ≤20
  • Freezing Point:
    ≥11.0 °C
  • Peroxide (as H₂O₂):
    ≤0.005 %
  • Residue after Evaporation:
    ≤0.0050 %
  • Titrable Acid (meq/g):
    ≤0.0016
  • Water (by KF, coulometric):
    ≤0.05 %

 

 

Frequently Bought Together