Order Entry
Austria
ContactUsLinkComponent
2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Irritant
2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Irritant
2-Propanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99.5 %
  • Carbonyl Compounds (max. 0.002% Propionaldehyde and max. 0.002% Acetone):
    passes test
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 5 ppm
  • Solubility in Water:
    passes test
  • Titrable Acid or Base (meq/g):
    max. 0.0001
  • Water (H2O):
    max. 0.05 %
  • at 210 nm:
    max. 1.00
  • at 220 nm:
    max. 0.40
  • at 230 nm:
    max. 0.20
  • at 245 nm:
    max. 0.08
  • at 260 nm:
    max. 0.04
  • at 275 nm:
    max. 0.03
  • at 300 nm:
    max. 0.02
  • at 330-400 nm:
    max. 0.01

Specification Test Results

Related Information

Frequently Bought Together