
Schiff reagent
Catalog # J62171.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
Density:1,1 g/cm³ (20 °C)
:GHS07
Storage Temperature:Refrigerator
MDL Number:MFCD00081985