Buffer, sodium acetate, 3 M aq, soln,, pH 7,0, RNAse free
Artikelnr. J61934.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- CAS No.:127-09-3
- Description:Sodium acetate, 3M aq, solution, pH 7,0, RNAse free
- MDL No.:MFCD00012459
- Formula weight:82.03
- Physical form:Liquid
- Molecular formula:C2H3NaO2