Order Entry
Austria
ContactUsLinkComponent
Calcium nitrate tetrahydrate 99.0-103.0%, crystals, AR® ACS, Macron Fine Chemicals™
Oxidizing
Corrosive
Calcium nitrate tetrahydrate 99.0-103.0%, crystals, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Oxidizing
Corrosive
Calcium nitrate tetrahydrate 99.0-103.0%, crystals, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 103.0%
  • For Laboratory,Research,or Manufacturing Use:
    5.0 - 7.0
  • ACS - Assay (Ca(NO₃)₂·4H₂O):
    99.0 - 103.0 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 7.0
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Nitrite (NO₂):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.002 %
  • ACS - Barium (Ba):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Magnesium (Mg):
    ≤0.05 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Sodium (Na):
    ≤0.01 %
  • ACS - Strontium (Sr):
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together