
4'-Chloro-3'-methylacetophenone ≥75% remainder 3'-chloro-4'-methylacetophenone, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37074-39-8
:
Formula:C₉H₉ClO
MDL Number:MFCD00045172
:168.62 g/mol
Boiling Pt:104…105 °C (0.4 mmHg)
Density:1.15
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together