Pig Desoxyribonuclease II Typ IV (from Spleen)
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:9025-64-3
- Species:Pig
- Enzyme name:Desoxyribonuclease II, Typ IV
- Enzyme synonyms:DNase II
Specifications
Frequently Bought Together