Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA13490000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:93-72-1
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid 100 µg/ml in acetonitrile
- Formula:C₉H₇Cl₃O₃
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid