Calcium perchlorate tetrahydrate, Sigma-Aldrich®
Catalog # SIAL401420-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
:
EINECS:236-768-0
CAS Number:15627-86-8
Formula:CaClO₄·4H₂O
MDL Number:MFCD00191895