Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Catalog # MAYBCD03352.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:670.28 g/mol
CAS Number:175136-62-6
Formula:C₂₄H₉F₁₈P
MDL Number:MFCD00042428