Potassium acetate 3 mol/l (3 N) in aqueous solution
Catalog # R011
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:98.14 g/mol
Storage Temperature:Ambient
EINECS:204-822-2
CAS Number:127-08-2
Formula:CH₃COOK
MDL Number:MFCD00012458