Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Phenanthrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, Phenanthrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20920000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Phenanthrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20920000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    85-01-8
  • Description:
    Phenanthrene 10 µg/ml in cyclohexane
  • Formula:
    C₁₄H₁₀
  • Description:
    Phenanthrene

Specifications

About this item