D-(+)-Cellobiose, Millipore®
Catalog # 219458-25
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:342.30 g/mol
Storage Temperature:Ambient
EINECS:208-436-5
CAS Number:528-50-7
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00136034