Pesticide reference standard, 1,3-Dichlorobenzene 100 µg/ml in isooctane, CIL
Catalog # PROOCILULM74311.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:541-73-1
- Description:1,3-Dichlorobenzene 100 µg/ml in isooctane
- Formula:C₆H₄Cl₂
- Description:1,3-Dichlorobenzene