1,1-Dimethyl-4-phenylpiperazinium iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:54-77-3
:
Formula:C₁₂H₁₉IN₂
MDL Number:MFCD00011976
:M-Clarity™ quality level = MQ200
EINECS:200-213-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together